Learn More
MadgeTech™ General-Purpose pH Probe
SDP
Catalog No. 15060053
Change view
Click to view available options
For Use With (Equipment):
pHTemp101 Datalogger
Catalog No. For Use With (Equipment)
15-060-053 pHTemp101 Datalogger
1 options
Catalog No. 15-060-053 Supplier MadgeTech™ Supplier No. PH1
Add to Cart
Add to Cart
For use with the pHTemp101 Datalogger
  • Features standard BNC connector
  • Clear, epoxy-bodied, gel-filled combination electrode measures the entire 0 to 14pH range
  • Soaker storage bottle included
Specifications
Type Probe, pH
Includes Soaker storage bottle
Material Epoxy
For Use With (Equipment) pHTemp101 Datalogger