Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MadgeTech™ VTMS™ Vaccine Temperature Datalogger System
SDP

Catalog No. 15060701
Change view
Click to view available options
Battery Life:
3 Years
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Life
15-060-701 3 Years
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-060-701 Supplier MadgeTech™ Supplier No. VTMS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

USB interface package (Cat. No. 15-060-036)

For data logging of temperature-sensitive vaccines or pharmaceuticals

In real-time chart recording mode, the VTMS can alert the user, via e-mail or text message if the refrigerator has failed, or if the temperature is outside of the nominal range. This allows users to take immediate actions to ensure vaccines and pharmaceuticals in storage are not destroyed. To prevent false alarms, the unit is connected to a Glycol bottle which provides temperature buffered readings.

  • Ideal for continuous measurement and data logging of temperature sensitive vaccines or pharmaceuticals in refrigerators, freezers, and coolers
  • System includes a N.I.S.T. traceable, TCTemp2000 datalogger with thermocouple, Glycol bottle and power supply
  • Data logger has convenient LCD to ensure temperature parameters are met, in real time
  • Current reading, minimum, maximum and average statistics can all be obtained through the display
General Specifications:
  • Memory: 131,071/channel
  • Reading Rate: 1 reading every 2 seconds to 1 every 24 hours
  • Baud Rate: 115,200
  • Cold Junction Compensation: Automatic, based on internal channel
  • Battery Life: 1 year, at 1 min. reading rate w/ display off; 30 days typical w/ continuous LCD and no backlight
  • Material: Black anodized aluminum
  • Dimensions: 4.8 x 3.3 x 1.25 in. (12.2 x 8.4 x 3.2cm)
  • Operating Environment: -20 to +60°C, 0 to 95% RH noncondensing
  • Required Interface Package: USB interface cable (Cat. No. 15-060-036)
Temperature:
  • Internal Channel Temperature Sensor: Semiconductor
  • Internal Channel Temperature Range: -20 to 60°C
  • Internal Channel Temperature Resolution: 0.1°C
  • Internal Channel Temperature Calibrated Accuracy: ±1.0°C
  • Remote Channel Temperature Sensor: Type K Thermocouple (included)

Vaccine temperature monitoring (refrigerators/freezers), Medical and pharmaceutical monitoring, Process verification and validation.

Compliance

CE approved.

Specifications

Type Vaccine Temperature Monitoring System
Data Memory 16,128/Channel
Calibration Digital Calibration
Battery Type 9 V Lithium
Battery Life 3 Years
Includes NIST traceable RFTCTemp2000A datalogger with thermocouple, Glycol bottle, power supply
Certifications/Compliance NIST
Material ABS Plastic
Model VTMS
Sensor Types Semiconductor
For Use With (Application) For the continuous measurement and data logging of temperature sensitive vaccines or pharmaceuticals in refrigerators, freezers and coolers
Humidity Range 0 to 95%
Temperature Resolution 0.1°C
Operating Temperature (Metric) -20°C to +60°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.