Learn More
Mopec Maestro Encore Safety Sash
SDP
Catalog No. 23987032 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Safety Sash, Left
Safety Sash, Right
Quantity:
48 Units
60 Units
72 Units
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
23-987-032 Safety Sash, Left 48 Units
23-987-033 Safety Sash, Left 60 Units
23-987-034 Safety Sash, Left 72 Units
23-987-035 Safety Sash, Right 48 Units
23-987-036 Safety Sash, Right 60 Units
23-987-037 Safety Sash, Right 72 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 23-987-032 Supplier Mopec Supplier No. AX124L048
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Enhance laboratory safety and workflow with the AX124 Safety Sash.
  • Designed to reduce exposure to formalin fumes and hazardous chemical spills and splashes
  • Engineered to work with PathCam imaging systems, Ensemble accessories, and formalin dispensing
  • Frameless door
  • Friction hinges
  • 304 stainless steel sheetmetal and hardware
  • Ideal for open formalin processing operations
  • Ensures maximum fume capture even in high traffic cross flow laboratory conditions
Specifications
Product Type Grossing Station
Material Stainless Steel (304)
For Use With (Equipment) Safety Sash, Left
Quantity 48 Units