Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113935 Supplier Invitrogen™ Supplier No. PA5113935
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVLEALSPPQ TSGLSPSRLS KSQGGEDESP LSDKCSRKTL FYLIATLNES For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

This gene encodes a protein that is similar to Maf1, a Saccharomyces cerevisiae protein highly conserved in eukaryotic cells. Yeast Maf1 is a negative effector of RNA polymerase III (Pol III). It responds to changes in the cellular environment and represses pol III transcription. Biochemical studies identified the initiation factor TFIIIB as a target for Maf1-dependent repression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MAF1
Gene Accession No. Q9D0U6
Gene Alias 1110068E11Rik; AU042856; homolog of yeast MAF1; MAF1; MAF1 homolog (S. cerevisiae); MAF1 homolog, negative regulator of RNA polymerase III; MAF1 negative regulator of RNA polymerase III; repressor of RNA polymerase III transcription MAF1 homolog
Gene Symbols MAF1
Host Species Rabbit
Immunogen Immunogen sequence: HVLEALSPPQ TSGLSPSRLS KSQGGEDESP LSDKCSRKTL FYLIATLNES (aa 54-103).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68877
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.