Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFB Recombinant Rabbit Monoclonal Antibody (BLR046F)
GREENER_CHOICE

Catalog No. PIMA544287
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544287 100 μL
PIMA544288 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544287 Supplier Invitrogen™ Supplier No. MA544287
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFB
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR046F
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.09% sodium azide; pH 8.2
Gene MAFB
Gene Accession No. Q9Y5Q3
Gene Alias b-maf; c-Maf; kr; Kreisler; Kreisler (maf-related) leucine zipper protein; Kreisler maf-related leucine zipper homolog; Kreisler maf-related leucine zipper homolog (b-maf); KRML; Krml1; LOW QUALITY PROTEIN: transcription factor MafB; maf; MAF bZIP transcription factor B; Maf1; MAFB; maf-B; MAFB/Kreisler basic region/leucine zipper transcription factor; MCTO; MGC43127; segmentation protein Kr; si:ch73-337g9.1; transcription factor Maf; Transcription factor Maf-1; transcription factor MafB; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog b (paralog b); v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B (avian); V-maf musculoaponeurotic fibrosarcoma oncogene homolog B
Gene Symbols MAFB
Host Species Rabbit
Immunogen A peptide representing a region between residues 1 and 50 of human MAF bZIP transcription factor B (MafB).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9935
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.