Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAFG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530086 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530086 Supplier Invitrogen™ Supplier No. PA530086
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, mouse muscle, rat muscle. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (92%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Globin gene expression is regulated through nuclear factor erythroid-2 (NFE2) elements located in enhancer-like locus control regions positioned many kb upstream of alpha- and beta-gene clusters (summarized by Blank et al., 1997 [PubMed 9166829]). NFE2 DNA-binding activity consists of a heterodimer containing a ubiquitous small Maf protein (MafF, MIM 604877; MafG; or MafK, MIM 600197) and the tissue-restricted protein p45 NFE2 (MIM 601490). Both subunits are members of the activator protein-1-like superfamily of basic leucine zipper (bZIP) proteins (see MIM 165160).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAFG
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene MAFG
Gene Accession No. O15525, O54790, Q76MX4
Gene Alias AA545192; avian musculoaponeurotic fibrosarcoma virus (v-maf) AS42 oncogene, protein G; basic leucine zipper transcription factor MafG; C630022N07Rik; hMAF; MAF bZIP transcription factor G; MAFG; MGC13090; MGC20149; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein G (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog G
Gene Symbols MAFG
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MAFG. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17134, 4097, 64188
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.