Description
- Magnetic separation of cells is achieved in minutes
- For negative cell selection, the undesired cells are retained on the walls of the tubes while the cells of interest remain in suspension
- For positive cell selection, the cells of interest are retained on the walls of the tubes while the undesired cells remain in suspension
Specifications
Specifications
| Product Line | R&D Systems |
| Quantity | Each |