Learn More
R&D Systems™ MagCellect Plus Human E-Cadherin+ Cell Isolation Kit

Description
A mononuclear cell suspension is first incubated with the Human E-Cadherin Biotinylated Antibody that targets the desired cells. MagCellect™ Streptavidin (SA) Ferrofluid is next added to the reaction that allows the SA coated nanoparticles to interact with the cells tagged with the monoclonal antibodies. The tube containing the cell suspension is then placed within a magnetic field. Magnetically tagged cells will migrate toward the magnet (desired cell fraction), leaving the untagged cells (unwanted cell population) in suspension to be harvested by aspiration while the tube remains in the magnetic field. The tube containing the magnetically selected (desired) cells is then removed from the magnet and the cells are resuspended in MagCellect™ Plus Buffer or tissue culture media. The enriched cell preparation is then available for a variety of applications including tissue culture, immune status monitoring and flow cytometry
Kit Contents:
- Human E-Cadherin Biotinylated Antibody (625 μL of Human E-Cadherin biotinylated antibody in a phosphate buffered solution containing BSA and preservative)
- Human E-Cadherin Alexo Fluor™ 647 Antibody (125 μL of Alexa Fluor™ 647-conjugated Goat anti-human E-Cadherin Detection antibody)
- Streptavidin Ferrofluid (1.25 mL of a solution containing BSA and preservatives)
- Plus 2 bottles (50 mL/bottle) of a 10X concentrated buffer
Specifications
Specifications
| Gene ID (Entrez) | 999 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.