Learn More
Description
Specifications
Specifications
| Conjugate | DyLight 550 |
| Specificity | This antibody binds human MAGEA3 and MAGEA6, but does not cross react with other members of the MAGE family like MAGE-A1, MAGE-A4 and MAGE-A10. |
| Target Species | Human |
| For Use With (Application) | Flow Cytometry |
| Gene ID (Entrez) | 4102 |
| Gene Symbol | MAGEA3 |
| Quantity | 0.1 mL |
| Research Discipline | Cancer |
| Storage Requirements | Store at 4C in the dark. |
| Synonym | Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
