Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAGEA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103303
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103303 Supplier Thermo Scientific Supplier No. PA5103303
Only null left
Add to Cart
Add to Cart

MAGEA9 Polyclonal Antibody

MAGEA9 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEA9
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAGEA9
Gene Accession No. P43362
Gene Alias Cancer/testis antigen 1.9; cancer/testis antigen family 1, member 9; CT1.9; MAGE family member A9; MAGE9; MAGE-9 antigen; MAGEA9; MAGEA9A; MAGEA9B; melanoma antigen family A, 9; melanoma antigen family A9; melanoma-associated antigen 9; MGC114427; similar to melanoma antigen family A, 5; uncharacterized protein LOC317297
Gene Symbols MAGEA9
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAGEA9; MAGEA9B(Accession P43362), corresponding to amino acid residues M1-S41.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4108
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.