Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAGEB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562654 Supplier Invitrogen™ Supplier No. PA562654
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAGSDERANS RDEEYPCSSE VSPSTESSCS NFINIKVGLL EQFLLY Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 30%.

MAGEB5 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAGEB5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAGEB5
Gene Accession No. Q9BZ81
Gene Alias 1700056A17Rik; Cancer/testis antigen 3.3; cancer/testis antigen family 3, member 3; CT3.3; MAGE family member B5; MAGE family testis and tumor-specific protein; MAGEB5; MAGE-B5; MAGE-B5 antigen; melanoma antigen family B, 5; melanoma antigen family B5; melanoma antigen, family B, 5; melanoma-associated antigen B5; mMage-b5
Gene Symbols MAGEB5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAGEB5. Recombinant protein control fragment (Product #RP-101579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.