Learn More
Invitrogen™ MAGEB5 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MAGEB5.
Specifications
Specifications
| Antigen | MAGEB5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MAGEB5 |
| Gene Accession No. | Q9BZ81 |
| Gene Alias | 1700056A17Rik; Cancer/testis antigen 3.3; cancer/testis antigen family 3, member 3; CT3.3; MAGE family member B5; MAGE family testis and tumor-specific protein; MAGEB5; MAGE-B5; MAGE-B5 antigen; melanoma antigen family B, 5; melanoma antigen family B5; melanoma antigen, family B, 5; melanoma-associated antigen B5; mMage-b5 |
| Gene Symbols | MAGEB5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.