Learn More
Research Products International Corp Magenta-GlcA 100 MG
SDP
Supplier:  Research Products International Corp B730500.1
Encompass_Preferred
Chromogenic substrate for B-D-Glucuronidase (GUS). Produces a magenta color for (GUS) gene detection.
Catalog No. 50-223-0379
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.