Learn More
Invitrogen™ MAGI2 Polyclonal Antibody

Description
Recommended positive controls: A431, H1299, HeLa, HepG2, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | MAGI2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | MAGI2 |
| Gene Accession No. | Q86UL8 |
| Gene Alias | activin receptor interacting protein 1; activin receptor-interacting protein 1; Acvri1; ACVRINP1; Acvrip1; AIP1; AIP-1; ARIP1; Atrophin-1-interacting protein 1; atrophin-1-interacting protein A; KIAA0705; MAGI2; MAGI-2; membrane associated guanylate kinase, WW and PDZ domain containing 2; membrane-associated guanylate kinase inverted 2; Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 2; mKIAA0705; Sscam; S-SCAM; Synaptic-scaffolding molecule |
| Gene Symbols | MAGI2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.