Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagicMedia™ E. coli Expression Medium, Powder

Catalog No. K6810
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
5 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K6810 1 L
K6815 5 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K6810 Supplier Invitrogen™ Supplier No. K6810
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Contains a proprietary Component A (in foil pouch powder format), and Component B (in buffer liquid format).

MagicMedia E. coli Expression Medium is specifically designed to dramatically increase the yield of recombinant proteins in T7-regulated E. coli expression systems without monitoring culture OD (optical density) or adding inducer. The proprietary MagicMedia formulation enables E. coli growth to reach culture densities 3- to 10-fold higher than those achieved with traditional LB + IPTG methods, allowing you to:

• Achieve 3–10 times higher protein yields (Figure 1) with much less hands-on time
• Eliminate time-consuming OD monitoring steps—simply inoculate, grow the culture overnight, and harvest (Figure 2)
• Reduce culture volume to simplify handling and scale-up processes
• Ideal for simultaneous expression of multiple clones for high-throughput applications

MagicMedia E. coli Expression Medium, powder, consists of a proprietary Component A (in foil pouch powder format) and Component B (in buffer liquid format).

MagicMedia E. coli Expression Medium is also available in a ready-to-use, pre-sterilized liquid format (Cat. No. K6803).

Order Info

Store Component A (foil pouches) and Component B buffer (liquid) at Room Temperature (15 - 30°C).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Component A (foil pouches) and Component B (liquid) at room temperature (15–30°C).
Form Powder, Liquid
Format Kit
Product Type Microbiology Media
Product Line MagicMedia
Quantity 1 L
Species Escherichia coli
Can I use a lower incubation temperature for MagicMedia Medium?

Yes, expression at 18 degrees C and 25 degrees C has been validated using the Dual Temperature expression protocol.

I am using a Champion pET vector with the BL21-AI cells. Can I use your MagicMedia Medium for expression?

Unfortunately, MagicMedia Medium will not work with the arabinose inducer. MagicMedia Medium needs an IPTG-inducible system along with an intact Lac operon.

What is MagicMedia Medium?

MagicMedia Medium is an E. coli expression medium. It produces 3-10 times higher protein yield, is compatible with T7 regulated E. coli vectors that contain a functional lac operon and BL21 (DE3) strains, is simple to use, and eliminates OD monitoring/induction steps.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.