Learn More
Julabo USA Inc MAGIO MS-1200FW R1270 EA 230V
SDP
Supplier:  Julabo USA Inc 9032728.N1.33
Encompass_Preferred
Water-Cooled Refrigerated/heating circulator, JULABO, MAGIO MS-1200FW 230V/60Hz, R1270 Refrigerant; -50…+200 °C; 2kW heater. Cooling: 1.26kW at 20C; 1.11kW at 0C; 640W at -20C, 390W @ -30C, 220W at -40C. Adjustable pressure/suction pump 16-31L/min, 3.5-13.3PSI. Color touchscreen UI with USB, RS232/RS485, Ethernet, MODBUS communication, USB data-logging; Pt100 port, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 97lbs
Catalog No. NC2750996
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov