Learn More
Julabo USA Inc Refrigerated/heating circulator, JULABO, MAGIO MS-450F, 230V/60Hz, R449A Refrigerant. -30…+200C. 2kW heater. Cooling 460W at 20C. Pump 16-31LPM 3.5-13 PSI. Touchscreen UI. USB, RS232/RS485, Ethernet. Pt100 port. Programmable. 3-4L bath tank
SDP
Supplier:  Julabo USA Inc 9032714.S1.33
Encompass_Preferred
Refrigerated/heating circulator, JULABO, MAGIO MS-450F 200-230V/50-60Hz, R449A Refrigerant; -30…+200 °C; 2kW heater. Cooling: 460W at 20C; 390W at 0C; 290W at -10C, 170W @ -20C, 70W at -30C. Adjustable pressure/suction pump 16-31L/min, 3.5-13.3PSI. Color touchscreen UI with USB, RS232/RS485, Ethernet, MODBUS communication, USB data-logging; Pt100 port, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 3-4L bath tank. 9.1 x 15.7 x 25.6 in, 64lbs
Catalog No. 50-224-6264
Only null left
Add to Cart
Request a Quote
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.