Learn More
Julabo USA Inc MAGIO MX-1800F R1270 EA 230V
SDP
Supplier:  Julabo USA Inc 9033751.N1.33
Encompass_Preferred
Refrigerated/heating circulator, JULABO, MAGIO MX-1800F 230V/60Hz, R1270 Refrigerant; -50…+200 °C; 3kW heater. Cooling: 1.8kW at 20C; 1.5kW at 0C; 700W at -20C, 400W @ -30C, 230W at -40C. Adjustable pressure/suction pump 16-31L/min, 3.5-13.3PSI. Color touchscreen UI with USB, RS232/RS485, Ethernet, MODBUS communication, USB data-logging; Pt100 port, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 5-7.5L bath tank. 15.7 x 19.7 x 33.9 in, 135lbs, with locking casters
Catalog No. NC2750997
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov