Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MagMAX™ Microbiome Lysis Solution

Catalog No. a42361
Encompass_Preferred
Change view
Click to view available options
Quantity:
800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42361 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42361 Supplier Applied Biosystems™ Supplier No. A42361
Add to Cart
Edge
Add to Cart

This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.

This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42357, and A42358). It is made available separately for applications that require more lysis solution than is provided in the kit.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.

Sample Type Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Quantity 800 mL
Shipping Condition Room Temperature
Target Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.