Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MagMAX™ Plant DNA Isolation Kit

Catalog No. A32580
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 Preps
384 Preps
96 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32580 384 Preps
A32549 96 Preps
A47156 1000 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A32580 Supplier Thermo Scientific™ Supplier No. A32580
Add to Cart
Add to Cart

The MagMAX™ Plant DNA Isolation Kit is designed for automated high-throughput or manual purification of DNA from a wide variety of plant species. The kit uses MagMAX magnetic bead technology, eliminating the need for phenol/chloroform extraction or alcohol precipitation.

The MagMAX™ Plant DNA Isolation Kit is designed for automated high-throughput or manual purification of DNA from a wide variety of plant species. The kit uses MagMAX magnetic bead technology, eliminating the need for phenol/chloroform extraction or alcohol precipitation. The procedures are optimized for the isolation of DNA from 10–100 mg plant samples. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, age of tissue sample, and storage conditions.

The MagMAX Plant DNA Isolation Kit relies on magnetic bead technology, enabling reproducible yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of the beads makes the technology ideal for automated high-throughput nucleic acid purification, as well as for manual purification for low sample throughput. The resulting high-quality DNA is free of proteins, nucleases, and other contaminants or inhibitors, and can be used in a wide range of downstream applications such as PCR, qPCR, or next-generation sequencing. See bar graph below for typical genomic DNA yields from various sources.

Features of the MagMAX Plant DNA Isolation Kit include:
• Versatile—suitable for most plant types and parts, including woody, polyphenol-rich, and lignified samples
• High DNA yields—compared to traditional CTAB method
• High quality—isolates high integrity DNA, free of inhibitors and RNA contamination
• Flexible—easily adjustable for high-throughput automation platforms and manual processing

Applications
Isolated DNA is ideal for use in common molecular biology procedures, including:
• qPCR
• next-generation sequencing

Benefits of MagMAX magnetic bead–based purification
The magnetic bead–based purification format of the kit allows for easy scaling of the number of samples processed either manually with a magnetic stand or with a magnetic particle processor such as the KingFisher™ Duo Prime or KingFisher™ Flex processors.

Magnetic beads offer many benefits compared to other technologies for isolating nucleic acids. Beads bind the RNA and DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of undigested tissue particulates clogging these items during the extraction process. This clogging issue is of particular concern with fibrous plant tissue samples that that are not fully lysed during the disruption and homogenization steps.

Optimized bead surface chemistry
The Dynabeads™ MyOne™ Carboxylic Acid utilized in the MagMAX Plant DNA Kit has an optimized surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads magnetic technology ideal for the recovery of the DNA targets found in a variety of plant samples. Once captured, the DNA is then eluted in 150 μL and ready for downstream applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 4 x 60 mL Lysis Buffer A, store at room temperature
• 4 x 8.4 mL Lysis Buffer B, store at room temperature
• 8 x 1.4 mL Rnase A, store at -5°C to -30°C
• 4 x 16.5 mL Precipitation Solution, store at room temperature
• 8 x 1.4 mL DNA Binding Beads, store at 2°C to 8°C
• 4 x 45 mL Wash Solution 1 Concentrate, store at room temperature
• 4 x 60 mL Wash Solution 2 Concentrate, store at room temperature
• 4 x 30 mL Elution Buffer, store at room temperature
Format Kit
Isolation Technology Magnetic Bead
Sample Type Plants
For Use With (Application) Real-Time PCR
High-throughput Compatibility High-throughput Compatible
Product Line MagMAX
Product Type Plant DNA Isolation Kit
Quantity 384 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less
What kind of Polyvinylpyrrolidone (PVP) do you recommend using with MagMAX Plant DNA and RNA isolation kits since there are different kinds available?

For the MagMAX Plant DNA Isolation Kit, we recommend using PVP M.W. 40,000, Cat. No. J62417.A1 (https://www.thermofisher.com/order/catalog/product/J62417.A1).

For the MagMAX Plant RNA Isolation Kit, we recommend using Plant RNA Isolation Aid, Cat. No. AM9690 (https://www.thermofisher.com/order/catalog/product/AM9690).

What is the advantage of adding polyvinyl pyrollidone (PVP) or dithiothreitol (DTT) to Lysis Buffer A of the MagMAX Plant DNA Isolation Kit?

PVP adsorbs polyphenols, removing them from the final eluate. DTT, a reducing agent, protects the DNA from degradation by preventing the oxidation of polyphenols. We generally recommend adding PVP for woody, lignified, or polyphenol samples (branches, twigs, needles, and wax-coated leaves). DTT can be helpful when working with seeds. Some seeds (e.g., seeds with a pesticide coat) do benefit from addition of PVP to Lysis Buffer A. If DTT supplementation to Lysis Buffer A was already attempted and has not helped, try again with PVP instead.

The manual for the MagMAX Plant Isolation Kit recommends using 150 µL of elution buffer, but can I use less than that?

If higher DNA concentration is needed, the elution volume can be reduced to 50 µL when using the KingFisher Flex instrument. However, we have not tested this and the script may have to be changed accordingly.

Where can I find the script for using the MagMAX Plant DNA Isolation Kit on the KingFisher Flex or KingFisher Duo instrument?

You can find the .bdz files on the product page (https://www.thermofisher.com/order/catalog/product/A32549?ICID=search-product).

What kind of plants have you tested with the MagMAX Plant DNA Isolation Kit?

Table 1 of the manual (https://tools.thermofisher.com/content/sfs/manuals/MAN0015954_MagMAX_Plant_DNA_Kit_UG.pdf) lists all the different plants we have tested with the kit and the approximate DNA yields to expect.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.