Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Magna ChIP™ A - Chromatin Immunoprecipitation Kit

Catalog No. 17610 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
22 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-610 22 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-610 Supplier MilliporeSigma™ Supplier No. 17610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Magnetic Protein A Beads
  • ChIP Dilution Buffer
  • Low Salt Wash Buffer
  • High Salt Wash Buffer
  • LiCl Wash Buffer
  • TE Buffer
  • Cell Lysis Buffer
  • Nuclear Lysis Buffer
  • ChIP Elution Buffer (w/o Proteinase K)
  • 10X Glycine
  • 10X PBS
  • Protease Inhibitor Cocktail II
  • Proteinase K
  • Spin Filters
  • Collection Tubes
  • Bind Reagent A
  • Wash Reagent B
  • Elution Reagent C

Requires

Magna Grip Rack 8 well

Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads

  • Faster: Magnetic protein A beads allow for the entire ChIP protocol to be done in as little as a day! All reagents to process your samples are included
  • Easier: Spin columns make DNA purification easier and more reliable
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at the temperatures indicated on the labels
For Use With (Application) 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads
Quantity 22 Assays
Stability Stable for 1 year from date of shipment when stored as directed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.