Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Magna ChIP™ HiSens Chromatin Immunoprecipitation Kit
Shop All MilliporeSigma ProductsDescription
Includes
- 10X Glycine
- Nuclei Isolation Buffer
- 10X PBS
- SCW Buffer (Sonication/ChIP/Wash)
- Magna ChIP Protein A/G Magnetic Beads
- Low Stringency IP Wash Buffer
- ChIP Elution Buffer
- Protease Inhibitor Cocktail
- Proteinase K Solution
Requires
Reagents
- Cells, stimulated or treated as desired
- Antibody of interest for chromatin immunoprecipitation
- 37% Formaldehyde
- qPCR reagents, such as SYBR™ Green Master Mix
- 2% Agarose gel
- 100 bp DNA Ladder
- 6X gel Loading buffer
- DNase- and RNase-free sterile H2O
- Robust ChIP from as few as 10,000 to as many as 1,000,000 cells
- Protocols and reagents for generation of chromatin from range of input sample type
- Superior performance with variety of isotypes of either polyclonal or monoclonal antibodies
- Specialized buffer and bead formulations; lower backgrounds and higher fold enrichment
- Single buffer system for sonication, chromatin IP, and wash
- Perform analysis of enrichment without additional purification after cross link reversal
- Compatible with all commonly used downstream analysis applications– qPCR, next generation sequencing, microarray
Specifications
Specifications
| Content And Storage | Store components at the temperatures indicated on the labels |
| For Use With (Application) | Chromatin Immunoprecipitation (ChIP), ChIP-seq, ChIP-chip |
| Quantity | 24 Assays |
| Stability | Kit components are stable for 1 year from date of shipment when stored as directed |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?