Learn More
Description
- Recombinant protein A covalently bound to magnetic beads
- These beads provide users a rapid, reproducible and efficient reagent for collecting immunocomplexes for chromatin immunoprecipitations (ChIP) assays
- Protein A magnetic beads significantly reduce the handling time and mechanical stress on target immunocomplexes
- Presentation: Liquid suspension. Supplied as magnetic bead slurry in phosphate buffered saline, pH 7.4, containing 0.01% Tween™-20 and 0.09% sodium azide.
- Application: Recombinant Protein A covalently bound to magnetic beads for use in chromatin immunoprecipitations (ChIP assays). These protein A beads provide users a more rapid, reproducible & efficient reagent for collecting immunocomplexes vs. agarose beads
- Store kit components at the temperatures indicated on the labels and insert until their expiration date
- Store HeLa Whole Cell Extract at -80°C. Avoid repeated freeze-thaw cycles. Stable at -80°C for 6 months
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Chromatin immunoprecipitations (ChIP assays) |
| Quantity | 50 Reactions |
| Sample Volume | 1 mL |
| Type | Magnetic Beads |
| Cell Line | HeLa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.