Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MagnaBot™ Magnetic Separation Devices

Catalog No. PRV8151 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Deep Well MagnaBot 96
MagnaBot 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRV8151 MagnaBot 96
PRV3031 Deep Well MagnaBot 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV8151 Supplier Promega Supplier No. V8151
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for high-throughput DNA purification when used with MagneSil™ paramagnetic particles

  • Designed for high-throughput bioseparation using magnetic particles
  • Compatible with many plates

Specifications

Content And Storage 15°C to 30°C
Isolation Technology Magnetic Bead
For Use With (Equipment) Magnesil™ paramagnetic particles
For Use With (Application) High-throughput bioseparation using Magnetic particles
Product Type MagnaBot 96
Quantity 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.