Learn More
ATCC Magnaporthe oryzae Couch strain Guy11, frozen ampoule (ATCC 201236)
Supplier:  ATCC 201236SPQ
Encompass
Magnaporthe oryzae Couch strain Guy11 is provided as a frozen ampoule containing viable fungal cells (spores and/or mycelia) suspended in cryoprotectant for laboratory research use only. The strain was isolated from rice (Oryza sativa) and is noted for virulence on susceptible rice cultivars; it requires ultra-low temperature storage and aseptic handling.
  • Frozen ampoule format containing viable spores and mycelia.
  • Strain designation: Guy11.
  • Isolation source: rice (Oryza sativa); origin French Guiana.
  • Mating type: MAT1-2.
  • Recommended medium: oatmeal agar.
  • Incubation: aerobic at 24-26°C.
  • Storage: -80°C or colder; liquid nitrogen recommended for long-term storage.
  • Intended for laboratory research only; not for diagnostic, therapeutic, or consumption use.
Catalog No. NC3788966