Learn More
United Scientific™ Magnaprobe with Magnetic Globe
SDPSCI_ED
Catalog No. S80016 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
K+
Catalog No. Grade
S80016 K+
1 options
Catalog No. S80016 Supplier United Scientific™ Supplier No. MGNGLBPRB
Add to Cart
Add to Cart
Magnaprobe with Magnetic Globe demonstrates Earth's invisible magnetic field.
  • The Earth acts like it has a huge bar magnet located at its center.
  • This set contains a globe that is 110 mm wide with a magnet contained along its axis.
  • The magnaprobe contains a small magnet that is allowed to rotate on two axes.
  • This set illustrates the path of the magnetic field along the surface of a globe.
  • The closer to the poles, the steeper the angle of the magnetic field.
  • This is useful in demonstrating the large magnetic field that exists on the Earth.
  • The activity guide contains information about the Earth's magnetic field, Van Allen belts, and the northern lights.
  • Detailed globe with countries.
Specifications
Grade K+
Product Type Magnetic Globe
Warning: Cancer and reproductive harm