Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Magne™ Protein A Beads, 20% Slurry

Catalog No. PRG8783 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8783 50 mL
PRG8781 1 mL
PRG8782 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG8783 Supplier Promega Supplier No. G8783
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Magne Protein G & Magne Protein A Beads are magnetic affinity beads with high specificity and capacity for purification of immunoglobulins from cell culture media, ascites and serum. Iron encapsulated in macroporous cellulose with low nonspecific binding.

  • High Specificity, High Capacity Magnetic Affinity Beads for Antibody Capture
  • Binding capacities of >25mg/mL depending on antibody species, isotype
  • Minimize purification losses and increase sample throughput with exceptional magnetic properties

Specifications

Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.