Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MagneSil™ Blood Genomic, Max Yield System

Catalog No. PRMD1360 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
96 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRMD1360 96 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRMD1360 Supplier Promega Supplier No. MD1360
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MagneSil™ Blood Genomic, Max Yield System provides automated high-throughput DNA purification on the Beckman Coulter Biomek FX using MagneSil™ Paramagnetic Particle technology.

  • Walkaway Automation of Genomic DNA Extraction
  • Purifies DNA from 96 samples of anti-coagulated whole blood in 1 1/2 hours
  • Average yield of 96 samples from normal healthy adults is ≥4μg
  • Validated automated methods available

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 96 Preps
Product Type DNA Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.