Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MagneSil™ BLUE for Wizard™ MagneSil™ Plasmid Purification System

Catalog No. PRA2201 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2201 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2201 Supplier Promega Supplier No. A2201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Wizard™ MagneSil™ Plasmid DNA Purification System provides a simple, reliable method for rapid isolation of plasmid DNA in a 96-well, high-throughput format.

  • Rapid Isolation of Plasmid DNA in 96-Well Format
  • Process multiple plates without user intervention
  • Validated automated methods available

Specifications

Content And Storage 15°C to 30°C
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.