Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MagneSil™ ONE, Fixed Yield Blood Genomic System

Catalog No. PRMD1370 Shop All Promega Corporation Products
Encompass
Catalog No. PRMD1370 Supplier Promega Supplier No. MD1370
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MagneSil™ ONE, Fixed Yield Blood Genomic System purifies 1μgof DNA (+/- 50%) from 60μL of anti-coagulated whole blood.

  • Automated Extraction of 1μg DNA from 60μL Blood
  • Process 96 samples in 1 hour with no hands-on time following robot setup
  • DNA suitable for use in PCR, multiplex PCR and SNP genotyping applications
  • Validated automated methods available

Specifications

Content And Storage 15°C to 30°C
Format Kit
No. of Reactions 1 x 96 Preps
Product Type Genomic DNA Isolation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.