Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium (0.006in. Thick Ribbon), Fisher Chemical

Catalog No. m81oz Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 oz.
Packaging:
Envelope
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M8-1OZ 1 oz. Envelope
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M8-1OZ
This item is not returnable. View return policy

0006 in. thick (0015cm)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Color Silver
pH 7
Melting Point 651°C
Boiling Point 1107°C
Atomic Weight 24.31
Quantity 1 oz.
Identification Pass Test
Packaging Envelope
Form Solid
Formula Weight 24.3
Chemical Name or Material Magnesium, 0.006 in. Thick Ribbon
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.