Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Educational Products Magnesium
SCI_ED

Catalog No. S75532 Shop All American Educational Products LLC Products
Change view
Click to view available options
Magnesium Metal Ribbon; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S75532 Magnesium Metal Ribbon; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S75532 Supplier American Educational Products Supplier No. 7807

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Silver through Gray
CAS 7439-95-4
Quantity 25 g
Molecular Formula Mg
Linear Formula Mg
MDL Number MFCD00085308
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES [Mg]
IUPAC Name magnesium
Molecular Weight (g/mol) 24.30
PubChem CID 5462224
ChEBI CHEBI:25107
Formula Weight 24.31
Packaging Metal Ribbon
Chemical Name or Material Magnesium
Show More Show Less
Safety handling Check Alert storage code Red
ShelfLife Indefinite
DOTInformation DOT Class 4.1, : Flammable Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.