Learn More
Magnesium AA Standard, 1000 ppm Mg in 3% HNO3, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 1,000ppm Mg |
| CAS Min % | 93.0, 4.89, 0.09 |
| CAS Max % | 97.0, 5.09, 0.1 |
| Equivalence | 1 mL = 1 mg Mg |
| Physical Form | Liquid |
| Color | Colorless to Yellow |
| pH | < 2 |
| Melting Point | Approximately 0°C |
| Type | Single-element Standard Solution |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.