Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Acetate Tetrahydrate, ≥99%, Reagent Grade, Solstice

Catalog No. 60048052 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-052 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-052 Supplier Solstice Supplier No. M06311KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
Synonym magnesium acetate tetrahydrate, cromosa, acetic acid, magnesium salt, tetrahydrate, unii-i01g0ejc3b, i01g0ejc3b, magnesium diacetate tetrahydrate, magnesium diethanoate tetrahydrate, acmc-20aklu, magnesiumacetatetetrahydrate, ksc497c8p
PubChem CID 134717
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 68°C
Color Colorless
pH 8 to 9 (at 20°C)
Physical Form Solid
Quantity 1 kg
Odor Vinegar-like
Packaging Poly bottle
Density 1.45g/cu.cm at 20°C
Linear Formula (CH3COO)2Mg · 4H2O
UN Number NONH for all modes of transport
Beilstein 3730605
Solubility Information Completely soluble in water
IUPAC Name magnesium(2+) diacetate tetrahydrate
Formula Weight 214.45g/mol
Percent Purity ≥99%
Chemical Name or Material Magnesium acetate tetrahydrate
Show More Show Less
RTECSNumber 205-554-9
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.