Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Magnesium Bromide Hexahydrate
SCI_ED

Catalog No. s25398 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25398 100 g Lab Grade
S25398A 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25398

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 13446-53-2
Molecular Formula Br2H12MgO6
Molecular Weight (g/mol) 292.20
MDL Number MFCD00149780
InChI Key LGLXXNHIGIJYQQ-UHFFFAOYSA-L
Synonym acmc-1bp1b, magnesium bromide-water 1/2/6, magnesium 2+ ion hexahydrate dibromide, magnesium 2+ hexahydrate dibromide
PubChem CID 17749097
IUPAC Name dibromomagnesium hexahydrate
SMILES O.O.O.O.O.O.Br[Mg]Br

Specifications

Color White
Quantity 100 g
Linear Formula MgBr2·6H2O
Formula Weight 292.25
Grade Laboratory
Packaging Lab Grade
Density 2.0g/cmsuperscript_3;
Chemical Name or Material Magnesium Bromide, Hexahydrate
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.