Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, 10 g/L in Mixed Alcohol Solvent, for determination of Salts in Crude Oil, Ricca Chemical
SDP

Catalog No. 446532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4465-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4465-32 Supplier Ricca Chemical Company Supplier No. 446532
Add to Cart
Add to Cart

Determination of Salts in Crude Oil

Chemical Identifiers

CAS 71-36-3 , 67-56-1 , 7732-18-5 , 7791-18-6
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Color Colorless
CAS Min % 46.0, 26.48, 23.14, 1.95
CAS Max % 48.0, 27.56, 24.09, 2.03
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Mixed Alcohol Solvent
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 10g/L
Chemical Name or Material Magnesium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.