Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride, Hexahydrate, Crystal, EP, BP, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18604042 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-042 6 x 500 g Poly Bottle
18-604-041 500 g Poly Bottle
18-604-043 2.5 kg Poly Pail
18-604-044 4 x 2.5 kg Poly Pail
18-604-045 12 kg Poly Pail
18-604-046 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-042 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA110500GMPLCS6
Add to Cart
Add to Cart

MA110, 7791-18-6, MgCl2·6H2O

Spectrum™ Chemical Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP is used as a nutritional supplement. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

pH 4.5-7.0
CAS Max % 100%
Quantity 6 x 500 g
Percent Purity 98 to 101%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.