Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium chloride hexahydrate, Solstice

Catalog No. 6003100 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-031-00 4 x 2.5 kg
600-30-96 100 g
60-031-01 500 g
600-30-98 1 kg
60-030-99 2.5 kg
60-031-02 6 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-031-00 Supplier Solstice Supplier No. M92724X2.5KG
Add to Cart
Add to Cart

ACS Reagent, 99.0-102.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Quantity 4 x 2.5 kg
Linear Formula MgCl2 · 6H2O
UN Number NONH for all modes of transport
Formula Weight 203.3g/mol
Percent Purity 99 to 102%
Chemical Name or Material Magnesium chloride hexahydrate
RTECSNumber OM2975000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.