Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Magnesium Chloride Hexahydrate, MP Biomedicals™

Catalog No. MP021914211 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914211 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021914211 Supplier MP Biomedicals™ Supplier No. 02191421.1
Add to Cart
Add to Cart

Can be generated from the acid leach solution of magnesite tailings. Magnesium Chloride Hexahydrate, MP Biomedicals has been used to prepare a buffer for the renaturation of denatured peptidoglycan hydrolytic enzymes. It is a potential phase change materi

Application include:

  • Potential phase change material (PCM) for storing thermal energy in solar cookers
  • To alter poly (vinyl alcohol) [PVA] films has been investigated
  • To reduce the flammability of cotton fabrics has been studied
  • To prepare renaturation buffer for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes
  • As a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones
TRUSTED_SUSTAINABILITY

Specifications

Grade ACS
Purity 99 to 102%
For Use With (Application) Magnesium chloride hexahydrate has been used to prepare renaturation buffer that is employed for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes. It may be used as a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones.
Content And Storage Store at Room Temperature (15–30°C)
Quantity 100 g
Molecular Weight (g/mol) 203.3g/mol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.