Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Chloride Hexahydrate Technical, Reagents

Catalog No. 01337190
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-190 500 g
01-337-191 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-190 Supplier Reagents Holdings Llc Supplier No. C2238500500A5
Add to Cart
Add to Cart

Magnesium Chloride Hexahydrate Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
Synonym Magnesium dichloride hexahydrate
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Boiling Point 1412°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Specific Gravity 1.56
Chemical Name or Material Magnesium Chloride Hexahydrate Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.