Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MgCl2 (magnesium chloride) (25 mM)

Catalog No. AB0359 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB-0359 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB-0359 Supplier Thermo Scientific™ Supplier No. AB0359
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ PCR reagents meet high quality standards to generate accurate results from even the most challenging molecular biology applications.

Thermo Scientific™ PCR reagents meet high quality standards to generate accurate results from even the most challenging molecular biology applications. Magnesium ions (Mg2+) are an important component of PCR reactions where they interact with the DNA template, dNTPs, and Taq DNA Polymerase.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Chloride
Concentration 25 mM
For Use With (Application) Standard PCR
Physical Form Liquid
Quantity 5 mL
Includes • 25 mM MgCl2, 5 mL

Store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.