Learn More
Abcam Magnesium Dodecyl Sulfate, Anionic Detergent, Solid, 25 g
Description
- Molecular weight: 551.1 g/mol
- Purity: >98%
- Form: Solid
- Product type: Anionic detergent
- Composition: 98% Magnesium dodecyl sulfate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Buffer | 98% Magnesium dodecyl sulfate |
| Formulation | 98% Magnesium dodecyl sulfate |
| Quantity | 25 g |
| For Use With (Application) | Sample preparation and detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.