Learn More
Magnesium fluoride, 99% (metals basis excluding Ca & Na), Ca+Na <1%
Description
Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1261°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 250 g |
| Assay Percent Range | 99%, Ca+Na <1% (Metals basis excluding Ca and Na) |
| Solubility Information | Sparingly soluble in water. Slightly soluble in dilute HNO3Insoluble in ethanol. |
| Formula Weight | 62.3 |
| Odor | Odorless |
| Refractive Index | 1.35-1.38 |
| Density | 3.148 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 231-995-1 |
| RTECSNumber | OM3325000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.