Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hexafluorosilicate hexahydrate, 98%

Catalog No. AAA131890C
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA131890C 10,000 g
AAA1318936 500 g
AAA131890E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA131890C Supplier Thermo Scientific Chemicals Supplier No. A131890C
Add to Cart

CAS: 18972-56-0 | MgSiF6·6H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18972-56-0
Molecular Formula MgSiF6·6H2O
MDL Number MFCD00150250

Specifications

Quantity 10,000 g
UN Number UN2853
Formula Weight 274.47 (166.39 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 1.79
Chemical Name or Material Magnesium hexafluorosilicate hexahydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: MAGNESIUM FLUOROSILICATE
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.