Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium hydrogen phosphate trihydrate, 97+%

Catalog No. p-7021798
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1427930 250 g
AAA142790B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1427930 Supplier Thermo Scientific Chemicals Supplier No. A1427930
Only null left
Add to Cart

CAS: 7782-75-4 | H7MgO7P | 174.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
PubChem CID 16204530
IUPAC Name magnesium(2+) trihydrate hydrogen phosphate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O

Specifications

Quantity 250 g
Merck Index 14,5681
Formula Weight 174.34 (120.28 Anhydrous)
Percent Purity 99%
Odor Odorless
Density 2.13
Chemical Name or Material Magnesium hydrogen phosphate trihydrate
EINECSNumber 231-823-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.