Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Magnesium Hydroxide
SCI_ED

Catalog No. S25406A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25406A 500 g Lab Grade
S25406 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25406A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
IUPAC Name magnesium(2+) dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

Color White
Quantity 500 g
Assay Percent Range >95%
Linear Formula Mg(OH)2
Formula Weight 58.33
Grade Laboratory
Packaging Lab Grade
Density 2.36g/mL
Chemical Name or Material Magnesium Hydroxide
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.