accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHMg

CAS RN 1309-42-8

IUPAC Name: magnesium(2+) dihydroxide
Synonyms: magnesia,|magnesium hydroxide|milk of magnesia|Mg(OH)2|magnesium hydrate|magnesiumhydroxid
Molecular Weight (g/mol): 58.32
Molecular Formula: H2MgO2
InChi Key: VTHJTEIRLNZDEV-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Mg++]
Quantity 
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (7)
Physical Form 
  • (7)
  • (6)
  • (2)
Grade 
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
Percent Purity 
  • (6)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (6)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (7)
  • (7)
  • (6)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (6)
  • (3)
  • (1)