Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene gycol BioUlt25KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 812552.5KG
Encompass
Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It helps in the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system which is required for the purification of biological materials. PEG also interacts with cell membrane thereby allowing cell fusion.
Catalog No. 50-289-8517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.