Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Hydroxide, Powder, 95.0-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18611428 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-428 500 g Poly Pail
18-611-427 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-428 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1048500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1309-42-8

Specifications

Physical Form Powder, Needles or Crystals
CAS Max % 1
Quantity 500 g
Loss on Drying 0.02
Loss on Ignition 30 to 33%
Packaging Poly Pail
Molecular Weight (g/mol) 58.32
Percent Purity 95 to 100.5%
Grade Reagent
Chemical Name or Material Magnesium Hydroxide, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.