Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Hydroxide, Powder, USP, 95-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604077 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-077 100 lb. Poly Bottle
18-604-074 500 g Poly Bottle
18-604-075 2.5 kg Poly Bottle
18-604-076 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-077 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MA115100LBBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MA115, 1309-42-8, Mg(OH)2

Spectrum™ Chemical Magnesium Hydroxide, Powder, USP is used to treat symptoms caused by too much stomach acid such as heartburn, upset stomach, or indigestion. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
IUPAC Name magnesium(2+) dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

CAS Max % 100%
Physical Form Powder, Needles or Crystals
Quantity 100 lb.
Percent Purity 95 to 100.5%
Grade USP
Loss on Drying 2%
Loss on Ignition 30 to 33%
Packaging Poly Bottle
Chemical Name or Material Magnesium Hydroxide, Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.